Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methanesulfonic acid
Synonym(s):
Methanesulfonic acid
Linear Formula:
CH
3
SO
3
H
CAS Number:
75-75-2
Molecular Weight:
96.11
EC Number:
200-898-6
Products
Product Category
buffers (1)
organic acids (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
471356
Methanesulfonic acid,
≥99.0%
Expand
View Pricing
8.06022
Methanesulfonic acid,
for synthesis
Expand
View Pricing
59510
Methanesulfonic acid,
suitable for HPLC, LiChropur
™
, ≥99.5% (T)
Expand
View Pricing