Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Metal Packed GC Column (fits Agilent, Configuration "A")
Products
Product Category
columns (7)
Brand
Supelco (7)
Manufacturer
Column Type
ID (mm)
Length (m)
Compatibility
Quality Segment
Column Material
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
12392-U
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase none, matrix 60/80 Carboxen-1000 support, L × O.D. × I.D. 15.0 ft (4.6 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12554
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase none, matrix 60/80 Tenax TA support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12804-U
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase 20% SP-2100 + 0.1% Carbowax 1500, matrix 100/120 SUPELCOPORT support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13014-U
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase 20% Sebaconitrile, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 2.0 ft (0.61 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12594
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase 3% SP
®
-1500, matrix 80/120 Carbopack B support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12484-U
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase none, matrix 100/120 Molecular Sieve 5A support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13121-U
Metal Packed GC Column (fits Agilent, Configuration "A"),
phase 10% Silar 5 CP, matrix 80/100 Chromosorb W HP support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing