
Meclizine hydrochloride

Synonym(s):
Meclizine dihydrochloride monohydrate, 1-([4-Chlorophenyl]phenylmethyl)-4-([3-methylphenyl]methyl)piperazine dihydrochloride monohydrate
Empirical Formula (Hill Notation):
C25H27ClN2 · 2HCl · H2O
CAS Number:
Molecular Weight:
481.89
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2