Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Manganin
®
- resistance alloy, Cu86Mn12Ni2
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 167
Page 1 of 9
Page 1 of 9
Compare
Product Number
Description
Pricing
GF30821622
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
powder, 500g, 50micron max. particle size, atomized condition
Expand
View Pricing
GF11063258
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
foil, 100 x 100mm, 0.04mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF57647560
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
powder, 100g, 50micron max. particle size, atomized condition
Expand
View Pricing
GF36511477
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF60781709
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
rod, 1000mm x 3.0mm diameter, annealed
Expand
View Pricing
GF37433549
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
insulated wire, 100m length, 0.05mm conductor diameter, 0.004mm insulation thickness, polyimide insulation
Expand
View Pricing
GF94703283
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
foil, 2m coil, 0.04mm thickness, annealed
Expand
View Pricing
GF53873501
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
insulated wire, 100m length, 0.1mm conductor diameter, 0.005mm insulation thickness, polyimide insulation
Expand
View Pricing
GF51167875
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
insulated wire, 20m length, 0.1mm conductor diameter, 0.005mm insulation thickness, polyimide insulation
Expand
View Pricing
GF13626659
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
insulated wire, 50m length, 0.05mm conductor diameter, 0.004mm insulation thickness, polyester insulation
Expand
View Pricing
GF16517145
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
rod, 100mm x 8mm diameter, annealed
Expand
View Pricing
GF26093246
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
insulated wire, 10m length, 0.1mm conductor diameter, 0.005mm insulation thickness, polyimide insulation
Expand
View Pricing
GF57862390
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
100m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF00682433
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
foil, 50 x 50mm, 0.02mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF88497792
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
50m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF83436396
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
foil, 0.1m coil, 100mm coil width, 0.635mm thickness, annealed
Expand
View Pricing
GF17513782
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
foil, 50 x 50mm, 0.5mm thickness, annealed
Expand
View Pricing
GF22655437
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
foil, 8mm disks, 0.005mm thickness
Expand
View Pricing
GF25694815
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
powder, 50g, 50micron max. particle size, atomized condition
Expand
View Pricing
GF13047139
Manganin
®
- resistance alloy, Cu86Mn12Ni2,
insulated wire, 5m length, 0.15mm conductor diameter, 0.015mm insulation thickness, polyimide insulation
Expand
View Pricing
Page 1 of 9
Page 1 of 9