Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Malachite Green oxalate salt
Synonym(s):
N
,
N
,
N
′,
N
′-Tetramethyl-4,4′-diaminotriphenylcarbenium oxalate, Basic Green 4
Empirical Formula (Hill Notation):
C
23
H
25
N
2
· C
2
HO
4
· 0.5C
2
H
2
O
4
CAS Number:
2437-29-8
Molecular Weight:
463.50
EC Number:
219-441-7
Products
Product Category
stains (3)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Physical Form
Product Line
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
M9015
Malachite Green oxalate salt,
Dye content, ≥90%, certified by the Biological Stain Commission, powder or crystals
Expand
View Pricing
32745
Malachite Green oxalate salt,
for microscopy, crystalline, S. No.: 754
Expand
View Pricing
M6880
Malachite Green oxalate salt,
Powder, crystals or chunks
Expand
View Pricing
46396
Malachite Green oxalate salt,
VETRANAL
®
, analytical standard
Expand
View Pricing