Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MakroPor, silicon membrane
Linear Formula:
Si
Products
Product Category
membranes (28)
Brand
Sigma-Aldrich (28)
Pore size (µm)
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
MP1501010
MakroPor, silicon membrane,
thickness 50 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP12001010
MakroPor, silicon membrane,
thickness 50 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP250105
MakroPor, silicon membrane,
thickness 200 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
MP84751010
MakroPor, silicon membrane,
thickness 475 μm, pore diameter 8 μm, pore size 12 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP83501010
MakroPor, silicon membrane,
thickness 350 μm, pore diameter 8 μm, pore size 12 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP250205
MakroPor, silicon membrane,
thickness 200 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
MP53501010
MakroPor, silicon membrane,
thickness 350 μm, pore diameter 5.5 μm, pore size 12 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP5350205
MakroPor, silicon membrane,
thickness 350 μm, pore diameter 5.5 μm, pore size 12 μm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
MP54751010
MakroPor, silicon membrane,
thickness 475 μm, pore diameter 5.5 μm, pore size 12 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP150105
MakroPor, silicon membrane,
thickness 50 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
MP8350105
MakroPor, silicon membrane,
thickness 350 μm, pore diameter 8 μm, pore size 12 μm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
MP12002010
MakroPor, silicon membrane,
thickness 50 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 20 mm × 20 mm, pkg of 10 pieces
Expand
View Pricing
MP5350105
MakroPor, silicon membrane,
thickness 350 μm, pore diameter 5.5 μm, pore size 12 μm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
MP2501010
MakroPor, silicon membrane,
thickness 200 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
MP25200201
MakroPor, silicon membrane,
thickness 200 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 20 mm × 20 mm, pkg of 10 pieces
Expand
View Pricing
MP150201
MakroPor, silicon membrane,
thickness 50 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 20 mm × 20 mm, pkg of 1 pieces
Expand
View Pricing
MP25200205
MakroPor, silicon membrane,
thickness 200 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
MP25200105
MakroPor, silicon membrane,
thickness 200 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
MP1200205
MakroPor, silicon membrane,
thickness 50 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
MP83502010
MakroPor, silicon membrane,
thickness 350 μm, pore diameter 8 μm, pore size 12 μm (interpore distance), size 20 mm × 20 mm, pkg of 10 pieces
Expand
View Pricing
Page 1 of 2
Page 1 of 2