Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Maize GMO Standard ERM-BF446
Synonym(s):
Maize MIR162, GMO Standard ERM-BF446
CAS Number:
9005-25-8
Products
Manufacturer
Agency
Application
Reference Material Type
Special Grade
certified reference material (5)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
ERMBF446B
Maize GMO Standard ERM-BF446,
100% (MIR162) Maize for the detection of GMO maize (MIR162) with PCR, ERM
®
, certified reference material
Expand
View Pricing
ERMBF446C
Maize GMO Standard ERM-BF446,
0.1% (MIR162) Maize for the detection of GMO maize (MIR162) with PCR, certified reference material, ERM
®
Expand
View Pricing
ERMBF446A
Maize GMO Standard ERM-BF446,
certified reference material, 0% (MIR162) Maize for the detection of GMO maize (MIR162) with PCR, ERM
®
Expand
View Pricing
ERMBF446D
Maize GMO Standard ERM-BF446,
1% (MIR162) Maize for the detection of GMO maize (MIR162) with PCR, ERM
®
, certified reference material
Expand
View Pricing
ERMBF446E
Maize GMO Standard ERM-BF446,
10% (MIR162) Maize for the detection of GMO maize (MIR162) with PCR, ERM
®
, certified reference material
Expand
View Pricing