Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Maize GMO Standard ERM-BF414
Synonym(s):
GMO Standard ERM-BF414, Maize GA21
Products
Manufacturer
Agency
Format
Chemical Composition
Application
Reference Material Type
Special Grade
certified reference material (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
ERMBF414F
Maize GMO Standard ERM-BF414,
ERM
®
, certified reference material, 4.3% GA21, for the detection of GMO maize (GA21) with PCR
Expand
View Pricing
ERMBF414A
Maize GMO Standard ERM-BF414,
ERM
®
, certified reference material, 0% GA21, for the detection of GMO maize (GA21) with PCR
Expand
View Pricing
ERMBF414D
Maize GMO Standard ERM-BF414,
ERM
®
, certified reference material, 1.0% GA21, for the detection of GMO maize (GA21) with PCR
Expand
View Pricing
ERMBF414B
Maize GMO Standard ERM-BF414,
ERM
®
, certified reference material, 0.1% GA21, for the detection of GMO maize (GA21) with PCR
Expand
View Pricing
ERMBF414C
Maize GMO Standard ERM-BF414,
ERM
®
, certified reference material, 0.5% GA21, for the detection of GMO maize (GA21) with PCR
Expand
View Pricing
ERMBF414E
Maize GMO Standard ERM-BF414,
ERM
®
, certified reference material, 1.7% GA21, for the detection of GMO maize (GA21) with PCR
Expand
View Pricing