Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnetic shielding alloy, Ni77Fe14Cu5Mo4
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 115
Page 1 of 6
Page 1 of 6
Compare
Product Number
Description
Pricing
GF31658362
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 0.5m coil, 0.25mm thickness, annealed
Expand
View Pricing
GF74485435
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 100 x 100mm, 0.1mm thickness, annealed
Expand
View Pricing
GF84537636
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 20m coil, 0.025mm thickness
Expand
View Pricing
GF25772104
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 1m coil, 0.25mm thickness, annealed
Expand
View Pricing
GF13942571
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, Ni 77%/Fe 14%/Cu 5%/Mo 4%, thickness 0.05 mm, length 0.5 m, coil width 150 mm
Expand
View Pricing
GF30132795
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 0.5m coil, 0.125mm thickness, annealed
Expand
View Pricing
GF24433848
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 100 x 100mm, 0.025mm thickness, light tight (lt)
Expand
View Pricing
GF23720161
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 2m coil, 0.05mm thickness
Expand
View Pricing
GF25915642
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 100 x 100mm, 0.25mm thickness, annealed
Expand
View Pricing
GF88126274
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, Ni77%/Fe14%/Cu 5%/Mo 4%, size 60 × 60 mm, thickness 0.15 mm, temper annealed
Expand
View Pricing
GF57229128
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
rod, 200mm x 6.35mm diameter
Expand
View Pricing
GF59921949
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
powder, 200g, 45micron max. particle size
Expand
View Pricing
GF35844419
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
powder, 100g, 45micron max. particle size
Expand
View Pricing
GF64964694
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 50 x 50mm, 0.25mm thickness, annealed
Expand
View Pricing
GF18965931
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 50 x 50mm, 0.1mm thickness, annealed
Expand
View Pricing
GF99316503
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 0.5m coil, 0.15mm thickness, annealed
Expand
View Pricing
GF74990728
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 100 x 100mm, 0.125mm thickness, annealed
Expand
View Pricing
GF07250543
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 150 x 150mm, 0.05mm thickness, light tight (lt)
Expand
View Pricing
GF36058002
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, 100 x 100mm, 0.05mm thickness, light tight (lt)
Expand
View Pricing
GF89629598
Magnetic shielding alloy, Ni77Fe14Cu5Mo4,
foil, Ni77%/Fe14%/Cu 5%/Mo 4%, thickness 0.15 mm, size 90 × 90 mm, temper annealed
Expand
View Pricing
Page 1 of 6
Page 1 of 6