Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts
Products
Brand
Sigma-Aldrich (9)
Color
Physical Form
Chemical Composition
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
LPC-605
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Soy PC 100 %
Expand
View Pricing
LPC-601
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Brain PC 100 %
Expand
View Pricing
LPC-602
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Egg PC 100 %
Expand
View Pricing
LPC-606
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Brain PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing
LPC-603
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Heart PC 100 %
Expand
View Pricing
LPC-609
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Liver PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing
LPC-604
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Liver PC 100 %
Expand
View Pricing
LPC-607
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Egg PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing
LPC-610
Lyophilized Liposomes - Natural Phosphatidylcholine (PC) Lipid Extracts,
Soy PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing