Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Luer-to-Threaded UTS connector
Synonym(s):
environmental, veterinary
Products
Product Category
connectors (9)
Brand
Sigma-Aldrich (1)
Manufacturer
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z117978
Luer-to-Threaded UTS connector,
male Luer lock to 10-32 standard thread, w/ wrench flats, nickel plated
Expand
View Pricing
Z261521
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 10-32 standard thread, nickel plated
Expand
View Pricing
Z261661
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 1/4-28 standard thread, 316 stainless steel
Expand
View Pricing
Z117986
Luer-to-Threaded UTS connector,
female Luer lock to 10-32 standard thread, nickel plated
Expand
View Pricing
Z261513
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 10-32 internal standard thread, nickel plated
Expand
View Pricing
Z261688
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 1/4-32 standard thread, 316 stainless steel
Expand
View Pricing
Z261653
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 10-32 standard thread, 316 stainless steel
Expand
View Pricing
Z261548
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 1/4-28 standard thread, nickel plated
Expand
View Pricing
Z261645
Luer-to-Threaded UTS connector,
Micro-Mate
®
female Luer to 10-32 internal standard thread, 316 stainless steel
Expand
View Pricing