Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Thyroxine
Synonym(s):
3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T
4
Linear Formula:
HOC
6
H
2
(I)
2
OC
6
H
2
(I)
2
CH
2
CH(NH
2
)CO
2
H
CAS Number:
51-48-9
Molecular Weight:
776.87
EC Number:
200-101-1
Products
Product Category
hormones (2)
proteins (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Optical Isomer
Sterility
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T2376
L-Thyroxine,
≥98% (HPLC)
Expand
View Pricing
T1775
L-Thyroxine,
powder, BioReagent, suitable for cell culture
Expand
View Pricing