Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Load flask for Aldrich
®
Kugelrohr apparatus
Products
Product Category
flasks (9)
Brand
Sigma-Aldrich (9)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z748226
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
Z748218
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
Z748196
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
Z748234
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 250 mL, joint: ST/NS 14/20
Expand
View Pricing
Z748285
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 2000 mL, joint: ST/NS 24/40
Expand
View Pricing
Z748277
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 1000 mL, joint: ST/NS 24/40
Expand
View Pricing
Z748250
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 250 mL, joint: ST/NS 24/40
Expand
View Pricing
Z748269
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 500 mL, joint: ST/NS 24/40
Expand
View Pricing
Z748242
Load flask for Aldrich
®
Kugelrohr apparatus,
capacity 500 mL, joint: ST/NS 14/20
Expand
View Pricing