
L-Lysine acetate

Synonym(s):
L-Lysine acetate salt, (S)-2,6-Diaminohexanoic acid
Empirical Formula (Hill Notation):
C6H14N2O2 · C2H4O2
CAS Number:
Molecular Weight:
206.24
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2