Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipase from
Candida rugosa
Synonym(s):
Triacylglycerol acylhydrolase, Triacylglycerol lipase, CCL
CAS Number:
9001-62-1
EC Number:
232-619-9
Products
Product Category
enzymes (4)
Brand
Sigma-Aldrich (4)
Biological Source
Color
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
L1754
Lipase from
Candida rugosa
,
Type VII, ≥700 unit/mg solid
Expand
View Pricing
L8525
Lipase from
Candida rugosa
,
lyophilized powder, ≥40,000 units/mg protein
Expand
View Pricing
62316
Lipase from
Candida rugosa
,
powder, yellow-brown, ≥2 U/mg
Expand
View Pricing
62302
Lipase from
Candida rugosa
,
lyophilized, powder (fine), 15-25 U/mg
Expand
View Pricing