Skip to Content
Products
Cart
0
AD
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
L-Glutamic Dehydrogenase from bovine liver
Synonym(s):
L-GLDH, L-Glutamate:NAD[P]
+
Oxidoreductase (deaminating), Glutamate Dehydrogenase from bovine liver
CAS Number:
9029-12-3
EC Number:
232-848-4
Products
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Tissue
Quality Segment
Optical Isomer
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G2626
L-Glutamic Dehydrogenase from bovine liver,
Type II, 50% glycerol solution, ≥35 units/mg protein
Expand
View Pricing
G7882
L-Glutamic Dehydrogenase from bovine liver,
Type III, lyophilized powder, ≥20 units/mg protein
Expand
View Pricing
G2501
L-Glutamic Dehydrogenase from bovine liver,
Type I, ammonium sulfate suspension, ≥40 units/mg protein
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Français - FR
Learn More