
L-Glutamic acid γ-(4-methoxy-β-naphthylamide)

Empirical Formula (Hill Notation):
C16H18N2O4
CAS Number:
Molecular Weight:
302.33
Products
Product Category
Brand
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing