Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lenz
®
Bistabil
®
separating funnel
Products
Product Category
funnels (15)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Z330639
Lenz
®
Bistabil
®
separating funnel,
capacity 250 mL, joint: ST/NS 29/32, PTFE key, graduated: no
Expand
View Pricing
Z330523
Lenz
®
Bistabil
®
separating funnel,
capacity 1,000 mL, joint: ST/NS 29/32, PTFE key, graduated
Expand
View Pricing
Z330663
Lenz
®
Bistabil
®
separating funnel,
capacity 2,000 mL, joint: ST/NS 29/32, PTFE key, graduated: no
Expand
View Pricing
Z330647
Lenz
®
Bistabil
®
separating funnel,
capacity 500 mL, joint: ST/NS 29/32, PTFE key, graduated: no
Expand
View Pricing
Z330507
Lenz
®
Bistabil
®
separating funnel,
capacity 250 mL, joint: ST/NS 29/32, PTFE key, graduated
Expand
View Pricing
Z330655
Lenz
®
Bistabil
®
separating funnel,
capacity 1,000 mL, joint: ST/NS 29/32, PTFE key, graduated: no
Expand
View Pricing
Z330485
Lenz
®
Bistabil
®
separating funnel,
capacity 50 mL, joint: ST/NS 19/26, PTFE key, graduated
Expand
View Pricing
Z330515
Lenz
®
Bistabil
®
separating funnel,
capacity 500 mL, joint: ST/NS 29/32, PTFE key, graduated
Expand
View Pricing
Z330612
Lenz
®
Bistabil
®
separating funnel,
capacity 50 mL, joint: ST/NS 19/26, PTFE key, graduated: no
Expand
View Pricing
Z330620
Lenz
®
Bistabil
®
separating funnel,
capacity 100 mL, joint: ST/NS 19/26, PTFE key, graduated: no
Expand
View Pricing
Z330469
Lenz
®
Bistabil
®
separating funnel,
capacity 1,000 mL, joint: ST/NS 29/32, glass key, graduated
Expand
View Pricing
Z330558
Lenz
®
Bistabil
®
separating funnel,
capacity 50 mL, joint: ST/NS 19/26, glass key, graduated: no
Expand
View Pricing
Z330426
Lenz
®
Bistabil
®
separating funnel,
capacity 50 mL, joint: ST/NS 19/26, glass key, graduated
Expand
View Pricing
Z330493
Lenz
®
Bistabil
®
separating funnel,
capacity 100 mL, joint: ST/NS 19/26, PTFE key, graduated
Expand
View Pricing
Z330531
Lenz
®
Bistabil
®
separating funnel,
capacity 2,000 mL, joint: ST/NS 29/32, PTFE key, graduated
Expand
View Pricing