Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Cysteine hydrochloride
Linear Formula:
HSCH
2
CH(NH
2
)COOH · HCl
CAS Number:
52-89-1
Molecular Weight:
157.62
EC Number:
200-157-7
Products
Product Category
amino acids (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Special Grade
Anhydrous (3)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C1276
L-Cysteine hydrochloride,
≥98% (TLC), suitable for UHPLC
Expand
View Pricing
30120
L-Cysteine hydrochloride,
≥99.0% (RT)
Expand
View Pricing
C7477
L-Cysteine hydrochloride,
≥98.0%, suitable for cell culture, BioReagent, non-animal source
Expand
View Pricing