Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laser Tough-Tags
™
Products
Product Category
labels (1)
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Length (in)
Color
Technique
Filter Products
Showing 1-20 of 38
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z366226
Laser Tough-Tags
™
,
white, size 1.28 in. × 0.5 in.
Expand
View Pricing
Z688363
Laser Tough-Tags
™
,
Microcentrifuge tube labels, white, L × W 0.94 in. × 0.5 in.
Expand
View Pricing
Z743647
Laser Tough-Tags
™
,
white, L × W 1.50 in. × 0.75 in., pkg of 1500
Expand
View Pricing
L5289
Laser Tough-Tags
™
,
L × W 23 mm × 8 mm
Expand
View Pricing
L5039
Laser Tough-Tags
™
,
L × W 1.0 in. × 1.0 in.
Expand
View Pricing
Z743629
Laser Tough-Tags
™
,
clear, L × W 1.50 in. × 0.50 in., pkg of 1600
Expand
View Pricing
Z743640
Laser Tough-Tags
™
,
orange, L × W 1.28 in. × 0.50 in., pkg of 2125
Expand
View Pricing
Z743652
Laser Tough-Tags
™
,
slenderedge, white, L × W 2.625 in. × 0.1875 in., pkg of 2475
Expand
View Pricing
Z743633
Laser Tough-Tags
™
,
for microscope slides, white, L × W 0.875 in. × 0.875 in., pkg of 2400
Expand
View Pricing
Z743628
Laser Tough-Tags
™
,
clear, L × W 1.50 in. × 0.75 in., pkg of 1200
Expand
View Pricing
Z743620
Laser Tough-Tags
™
,
assorted colors, L × W 1.28 in. × 0.50 in., pkg of 2125
Expand
View Pricing
Z743632
Laser Tough-Tags
™
,
for microscope slides, assorted colors, L × W 0.875 in. × 0.875 in., pkg of 2400
Expand
View Pricing
Z743626
Laser Tough-Tags
™
,
blue, L × W 1.50 in. × 0.75 in., pkg of 1500
Expand
View Pricing
Z743641
Laser Tough-Tags
™
,
orange, L × W 1.50 in. × 0.75 in., pkg of 1500
Expand
View Pricing
Z743621
Laser Tough-Tags
™
,
assorted colors, L × W 1.50 in. × 0.75 in., pkg of 1500
Expand
View Pricing
Z743650
Laser Tough-Tags
™
,
yellow, L × W 1.28 in. × 0.50 in., pkg of 2125
Expand
View Pricing
Z744118
Laser Tough-Tags
™
,
white, W × L 2.5 in. × 0.5 in., pack of 1500 ea (60 labels per sheet)
Expand
View Pricing
Z743622
Laser Tough-Tags
™
,
blue, L × W 0.875 in. × 0.875 in., pkg of 2400
Expand
View Pricing
Z743644
Laser Tough-Tags
™
,
red, L × W 1.28 in. × 0.50 in., pkg of 2125
Expand
View Pricing
Z743637
Laser Tough-Tags
™
,
green, L × W 1.50 in. × 0.75 in., pkg of 1500
Expand
View Pricing
Page 1 of 2
Page 1 of 2