Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laser Tough-Spots
®
Products
Brand
Sigma-Aldrich (10)
Color
Manufacturer
Color
Package Size
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
T1316
Laser Tough-Spots
®
,
diam. 3/8 in., white
Expand
View Pricing
Z743584
Laser Tough-Spots
®
,
diam. 3/8 in., assorted colors, pack of 3840 (192 labels per sheet)
Expand
View Pricing
Z743582
Laser Tough-Spots
®
,
diam. 1/2 in., white, pack of 3840 (192 labels per sheet)
Expand
View Pricing
Z743583
Laser Tough-Spots
®
,
diam. 1/2 in., assorted colors, pack of 3840 (192 labels per sheet)
Expand
View Pricing
Z743586
Laser Tough-Spots
®
,
diam. 7/16 in., white, pack of 4680 (234 labels per sheet)
Expand
View Pricing
L5539
Laser Tough-Spots
®
,
diam. 3/8 in., green
Expand
View Pricing
L5789
Laser Tough-Spots
®
,
diam. 3/8 in., red
Expand
View Pricing
L5414
Laser Tough-Spots
®
,
diam. 3/8 in., blue
Expand
View Pricing
L6789
Laser Tough-Spots
®
,
diam. 1 in., white
Expand
View Pricing
L6164
Laser Tough-Spots
®
,
diam. 1/2 in., green
Expand
View Pricing
L5914
Laser Tough-Spots
®
,
diam. 3/8 in., yellow
Expand
View Pricing
L6539
Laser Tough-Spots
®
,
diam. 1/2 in., yellow
Expand
View Pricing
L6664
Laser Tough-Spots
®
,
diam. 3/4 in., white
Expand
View Pricing
L5664
Laser Tough-Spots
®
,
diam. 3/8 in., orange
Expand
View Pricing
L6039
Laser Tough-Spots
®
,
diam. 1/2 in., blue
Expand
View Pricing
L6289
Laser Tough-Spots
®
,
diam. 1/2 in., orange
Expand
View Pricing
L6414
Laser Tough-Spots
®
,
diam. 1/2 in., red
Expand
View Pricing