Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laser Cryo-Tags
®
sidewall
Products
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z743431
Laser Cryo-Tags
®
sidewall,
assorted colors, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Expand
View Pricing
Z743433
Laser Cryo-Tags
®
sidewall,
green, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Expand
View Pricing
Z743436
Laser Cryo-Tags
®
sidewall,
yellow, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Expand
View Pricing
Z743432
Laser Cryo-Tags
®
sidewall,
blue, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Expand
View Pricing
L4662
Laser Cryo-Tags
®
sidewall,
white, L × W 0.5 in. × 0.25 in.
Expand
View Pricing
Z743434
Laser Cryo-Tags
®
sidewall,
orange, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Expand
View Pricing
Z743435
Laser Cryo-Tags
®
sidewall,
red, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Expand
View Pricing