Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lanthanum(III) oxide
Synonym(s):
Lanthanum(III) oxide
Linear Formula:
La
2
O
3
CAS Number:
1312-81-8
Molecular Weight:
325.81
EC Number:
215-200-5
Products
Brand
Sigma-Aldrich (4)
Supelco (3)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
L4000
Lanthanum(III) oxide,
≥99.9%
Expand
View Pricing
199923
Lanthanum(III) oxide,
99.99% trace metals basis
Expand
View Pricing
289205
Lanthanum(III) oxide,
suitable for AAS, ≥99.9%
Expand
View Pricing
1.12220
Lanthanum(III) oxide,
EMPLURA
®
Expand
View Pricing
1.10982
Lanthanum(III) oxide,
for atomic absorption spectroscopy
Expand
View Pricing
634271
Lanthanum(III) oxide,
nanopowder, <100 nm particle size (TEM), 99% trace metals basis
Expand
View Pricing
203556
Lanthanum(III) oxide,
99.999% trace metals basis
Expand
View Pricing