Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KT5823
Synonym(s):
KT5823
Empirical Formula (Hill Notation):
C
29
H
25
N
3
O
5
CAS Number:
126643-37-6
Molecular Weight:
495.53
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
420321
KT5823,
Highly specific, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase G (K
i
= 234 nM).
Expand
View Pricing
K1388
KT5823,
≥85% (HPLC), lyophilized powder
Expand
View Pricing