Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KIMBLE
®
disposable screw thread culture tube with white marking spot
Products
Product Category
tubes (6)
Diameter (mm)
Length (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
DWK73750-16100
KIMBLE
®
disposable screw thread culture tube with white marking spot,
round bottom borosilicate glass, tube diam. × L (16 mm) (100 mm), non-sterile
Expand
View Pricing
DWK73750-13100
KIMBLE
®
disposable screw thread culture tube with white marking spot,
round bottom borosilicate glass, tube diam. × L (13 mm) (100 mm), non-sterile
Expand
View Pricing
DWK73750-16125
KIMBLE
®
disposable screw thread culture tube with white marking spot,
round bottom borosilicate glass, tube diam. × L (16 mm) (125 mm), non-sterile
Expand
View Pricing
DWK73750-20150
KIMBLE
®
disposable screw thread culture tube with white marking spot,
round bottom borosilicate glass, tube diam. × L (20 mm) (150 mm), non-sterile
Expand
View Pricing
DWK73750-20125
KIMBLE
®
disposable screw thread culture tube with white marking spot,
round bottom borosilicate glass, tube diam. × L (20 mm) (125 mm), non-sterile
Expand
View Pricing
DWK73750-16150
KIMBLE
®
disposable screw thread culture tube with white marking spot,
round bottom borosilicate glass, tube diam. × L (16 mm) (105 mm), non-sterile
Expand
View Pricing