Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KIMBLE
®
centrifuge tube
Products
Product Category
tubes (7)
Capacity (mL)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
DWK73785-15
KIMBLE
®
centrifuge tube,
with screw-top finish, conical bottom clear glass, capacity (15 mL), without cap, non-sterile
Expand
View Pricing
DWK73785-10
KIMBLE
®
centrifuge tube,
with screw-top finish, conical bottom clear glass, capacity (10 mL), without cap, non-sterile
Expand
View Pricing
DWK73785-50
KIMBLE
®
centrifuge tube,
with screw-top finish, conical bottom clear glass, capacity (50 mL), without cap, non-sterile
Expand
View Pricing
DWK73790-15
KIMBLE
®
centrifuge tube,
with plain rim finish, conical bottom clear glass, capacity (15 mL), without cap, non-sterile
Expand
View Pricing
DWK73790-50
KIMBLE
®
centrifuge tube,
with plain rim finish, conical bottom clear glass, capacity (50 mL), without cap, non-sterile
Expand
View Pricing
DWK73790-10
KIMBLE
®
centrifuge tube,
with plain rim finish, conical bottom clear glass, capacity (10 mL), without cap, non-sterile
Expand
View Pricing
DWK73790-5
KIMBLE
®
centrifuge tube,
with plain rim finish, conical bottom clear glass, capacity (5 mL), without cap, non-sterile
Expand
View Pricing