Skip to Content
Merck
Ketanserin (+)-tartrate salt ≥97%, solid

Ketanserin (+)-tartrate salt

Ketanserin (+)-tartrate salt ≥97%, solid
Synonym(s):
3-(2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl)-2,4(1H,3H)-quinazolinedione (+)-tartrate salt, R 41468 (+)-tartrate salt
Empirical Formula (Hill Notation):
C22H22FN3O3 · C4H6O6
CAS Number:
Molecular Weight:
545.51
EC Number:

Products

Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing