Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern EMB series precision balances
Products
Product Category
balances (14)
Manufacturer
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
Z742801
Kern EMB series precision balances,
Kern EMB 2000-2, weighing capacity 2000 g, accuracy: 0.01 g, precision: ±0.05 g
Expand
View Pricing
Z742802
Kern EMB series precision balances,
Kern EMB 2200-0, weighing capacity 2200 g, accuracy: 1.0 g, precision: ±2.0 g
Expand
View Pricing
Z742785
Kern EMB series precision balances,
Kern EMB 500-1, weighing capacity 500 g, accuracy: 0.1 g, precision: ±0.2 g
Expand
View Pricing
Z742793
Kern EMB series precision balances,
Kern EMB 100-3, weighing capacity 100 g, accuracy: 0.001 g, precision: ±0.005 g
Expand
View Pricing
Z742794
Kern EMB series precision balances,
Kern EMB 200-2, weighing capacity 200 g, accuracy: 0.01 g, precision: ±0.02 g
Expand
View Pricing
Z742795
Kern EMB series precision balances,
Kern EMB 200-3, weighing capacity 200 g, accuracy: 0.001 g, precision: ±0.005 g
Expand
View Pricing
Z742804
Kern EMB series precision balances,
Kern EMB 5.2K5, weighing capacity 5000 g, accuracy: 5.0 g, precision: ±10.0 g
Expand
View Pricing
Z742800
Kern EMB series precision balances,
Kern EMB 1200-1, weighing capacity 1200 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing
Z742799
Kern EMB series precision balances,
Kern EMB 1000-2, weighing capacity 1000 g, accuracy: 0.01 g, precision: ±0.05 g
Expand
View Pricing
Z742798
Kern EMB series precision balances,
Kern EMB 600-2, weighing capacity 600 g, accuracy: 0.01 g, precision: ±0.03 g
Expand
View Pricing
Z742782
Kern EMB series precision balances,
Kern EMB 5.2K1, weighing capacity 5200 g, accuracy: 1.0 g, precision: ±3.0 g
Expand
View Pricing
Z742796
Kern EMB series precision balances,
Kern EMB 500-1BE, weighing capacity 500 g, accuracy: 0.1 g, precision: ±0.2 g, black Balance
Expand
View Pricing
Z742803
Kern EMB series precision balances,
Kern EMB 3000-1, weighing capacity 3000 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing
Z742806
Kern EMB series precision balances,
Kern EMB 6000-1, weighing capacity 6000 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing