Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern EG series Toploader balances
Products
Product Category
balances (10)
Manufacturer
Diameter (mm)
Height (mm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z662968
Kern EG series Toploader balances,
Kern EG 420-3NM, weighing capacity 420 g, resolution: 0.001 g, linearity: ±0.003 g
Expand
View Pricing
Z662933
Kern EG series Toploader balances,
Kern EG220-3NM, weighing capacity 220 g, resolution: 0.001 g, linearity: ±0.002 g
Expand
View Pricing
Z662976
Kern EG series Toploader balances,
Kern EG 420-3NM + 963-127, weighing capacity 420 g, resolution: 0.001 g, linearity: ±0.003 g, DKD Calibration Certificate included
Expand
View Pricing
Z662941
Kern EG series Toploader balances,
Kern EG220-3NM + 963-127, weighing capacity 220 g, resolution: 0.001 g, linearity: ±0.002 g, DKD Calibration Certificate included
Expand
View Pricing
Z742765
Kern EG series Toploader balances,
Kern EG 4200-2NM+YKA-03, weighing capacity 4200 g
Expand
View Pricing
Z662992
Kern EG series Toploader balances,
Kern EG 620-3NM + 963-121, weighing capacity 620 g, resolution: 0.001 g, linearity: ±0.004 g, DKD Calibration Certificate included
Expand
View Pricing
Z742763
Kern EG series Toploader balances,
Kern EG 2200-2NM+963-127+YKA-03, weighing capacity 2200 g,
DKD Calibration Certificate included
Expand
View Pricing
Z662984
Kern EG series Toploader balances,
Kern EG 620-3NM, weighing capacity 620 g, resolution: 0.001 g, linearity: ±0.004 g
Expand
View Pricing
Z742761
Kern EG series Toploader balances,
Kern EG 2200-2NM+YKA-03, weighing capacity 2200 g
Expand
View Pricing
Z742768
Kern EG series Toploader balances,
Kern EG 4200-2NM+963-127+YKA-03, weighing capacity 4200 g,
DKD Calibration Certificate included
Expand
View Pricing