Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern 572 series precision balances
Products
Product Category
balances (12)
Manufacturer
Diameter (mm)
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z742894
Kern 572 series precision balances,
Kern 572-49, weighing capacity 16000 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing
Z742891
Kern 572 series precision balances,
Kern 572-39, weighing capacity 4200 g, accuracy: 0.01 g, precision: ±0.05 g
Expand
View Pricing
Z742895
Kern 572 series precision balances,
Kern 572-55, weighing capacity 20000 g, accuracy: 0.05 g, precision: ±0.25 g
Expand
View Pricing
Z742887
Kern 572 series precision balances,
Kern 572-32, weighing capacity 420 g, accuracy: 0.001 g, precision: ±0.005 g
Expand
View Pricing
Z742896
Kern 572 series precision balances,
Kern 572-57, weighing capacity 24000 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing
Z742893
Kern 572 series precision balances,
Kern 572-45, weighing capacity 12000 g, accuracy: 0.05 g, precision: ±0.15 g
Expand
View Pricing
Z742890
Kern 572 series precision balances,
Kern 572-37, weighing capacity 3000 g, accuracy: 0.01 g, precision: ±0.05 g
Expand
View Pricing
Z742892
Kern 572 series precision balances,
Kern 572-43, weighing capacity 10000 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing
Z742889
Kern 572 series precision balances,
Kern 572-35, weighing capacity 2400 g, accuracy: 0.01 g, precision: ±0.03 g
Expand
View Pricing
Z742885
Kern 572 series precision balances,
Kern 572-30, weighing capacity 240 g, accuracy: 0.001 g, precision: ±0.003 g
Expand
View Pricing
Z742886
Kern 572 series precision balances,
Kern 572-31, weighing capacity 300 g, accuracy: 0.001 g, precision: ±0.005 g
Expand
View Pricing
Z742888
Kern 572 series precision balances,
Kern 572-33, weighing capacity 1600 g, accuracy: 0.01 g, precision: ±0.03 g
Expand
View Pricing