Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isopropyl β-D-1-thiogalactopyranoside
Synonym(s):
IPTG, Isopropyl β-D-thiogalactoside
Empirical Formula (Hill Notation):
C
9
H
18
O
5
S
CAS Number:
367-93-1
Molecular Weight:
238.30
EC Number:
206-703-0
Products
Product Category
carbohydrates (1)
detergents (1)
monosaccharides (1)
Brand
SAFC (1)
Sigma-Aldrich (1)
Biological Source
Color
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
PHG0010
Isopropyl β-D-1-thiogalactopyranoside,
Expand
View Pricing
I5502
Isopropyl β-D-1-thiogalactopyranoside,
≥99% (TLC)
Expand
View Pricing