Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isomaltose
Synonym(s):
6-O-α-D-Glucopyranosyl-D-glucose
Empirical Formula (Hill Notation):
C
12
H
22
O
11
CAS Number:
499-40-1
Molecular Weight:
342.30
EC Number:
207-879-1
Products
Product Category
carbohydrates (2)
disaccharides (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
I7253
Isomaltose,
~98% (TLC)
Expand
View Pricing
05389
Isomaltose,
analytical standard
Expand
View Pricing