Skip to Content
Products
Cart
0
AD
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Isoamyl alcohol
Synonym(s):
3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol
Linear Formula:
(CH
3
)
2
CHCH
2
CH
2
OH
CAS Number:
123-51-3
Molecular Weight:
88.15
EC Number:
204-633-5
Products
Product Category
aroma chemicals (2)
solvents (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Food Grade (2)
Halal (2)
Kosher (2)
Fragrance grade (1)
Natural (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
W205702
Isoamyl alcohol,
≥98%, FG
Expand
View Pricing
1.00979
Isoamyl alcohol,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
8.18969
Isoamyl alcohol,
for synthesis
Expand
View Pricing
W205710
Isoamyl alcohol,
natural, ≥98%, FG
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Français - FR
Learn More