Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron oxide (II,III), nanoparticles
Products
Brand
Sigma-Aldrich (12)
Physical Form
Quality Segment
Functional Group
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
900062
Iron oxide (II,III), nanoparticles,
30 nm avg. part. size (TEM), 5 mg/mL in H
2
O
Expand
View Pricing
900026
Iron oxide (II,III), nanoparticles,
15 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL in H
2
O
Expand
View Pricing
900042
Iron oxide (II,III), nanoparticles,
25 nm avg. part. size (TEM), 5 mg/mL in H
2
O
Expand
View Pricing
900043
Iron oxide (II,III), nanoparticles,
15 nm avg. part. size (TEM), 5 mg/mL in H
2
O
Expand
View Pricing
900082
Iron oxide (II,III), nanoparticles,
5 nm avg. part. size (TEM), 5 mg/mL in chloroform
Expand
View Pricing
900091
Iron oxide (II,III), nanoparticles,
10 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H
2
O
Expand
View Pricing
900084
Iron oxide (II,III), nanoparticles,
10 nm avg. part. size (TEM), 5 mg/mL in chloroform
Expand
View Pricing
900148
Iron oxide (II,III), nanoparticles,
30 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H
2
O
Expand
View Pricing
900064
Iron oxide (II,III), nanoparticles,
25 nm avg. part. size (TEM), 5 mg/mL in toluene
Expand
View Pricing
900200
Iron oxide (II,III), nanoparticles,
15 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL in H
2
O
Expand
View Pricing
900088
Iron oxide (II,III), nanoparticles,
20 nm avg. part. size (TEM), 5 mg/mL in chloroform
Expand
View Pricing
900092
Iron oxide (II,III), nanoparticles,
15 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H
2
O
Expand
View Pricing