Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron-Nickel alloy, Fe55Ni45
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
GF87951123
Iron-Nickel alloy, Fe55Ni45,
foil, 100 x 100mm, 0.1mm thickness
Expand
View Pricing
GF69503609
Iron-Nickel alloy, Fe55Ni45,
foil, 50 x 50mm, 0.1mm thickness
Expand
View Pricing
GF57356028
Iron-Nickel alloy, Fe55Ni45,
foil, 25 x 25mm, 0.1mm thickness
Expand
View Pricing
GF93890649
Iron-Nickel alloy, Fe55Ni45,
foil, 150 x 150mm, 0.1mm thickness
Expand
View Pricing
GF00479189
Iron-Nickel alloy, Fe55Ni45,
200mm rod, 13.5mm diameter
Expand
View Pricing
GF02142795
Iron-Nickel alloy, Fe55Ni45,
200mm rod, 6.0mm diameter
Expand
View Pricing
GF68817145
Iron-Nickel alloy, Fe55Ni45,
100mm rod, 6.0mm diameter
Expand
View Pricing
GF33126061
Iron-Nickel alloy, Fe55Ni45,
foil, 8mm disks, 0.1mm thickness
Expand
View Pricing
GF67849835
Iron-Nickel alloy, Fe55Ni45,
500mm rod, 6.0mm diameter
Expand
View Pricing
GF09350535
Iron-Nickel alloy, Fe55Ni45,
foil, 50mm disks, 0.1mm thickness
Expand
View Pricing
GF41065006
Iron-Nickel alloy, Fe55Ni45,
foil, 6mm disks, 0.1mm thickness
Expand
View Pricing
GF40758026
Iron-Nickel alloy, Fe55Ni45,
foil, 10mm disks, 0.1mm thickness
Expand
View Pricing
GF70728521
Iron-Nickel alloy, Fe55Ni45,
foil, 4mm disks, 0.1mm thickness
Expand
View Pricing
GF59898199
Iron-Nickel alloy, Fe55Ni45,
500mm rod, 13.5mm diameter
Expand
View Pricing
GF53603838
Iron-Nickel alloy, Fe55Ni45,
100mm rod, 13.5mm diameter
Expand
View Pricing
GF36330487
Iron-Nickel alloy, Fe55Ni45,
foil, 1m coil, 0.1mm thickness
Expand
View Pricing
GF44074807
Iron-Nickel alloy, Fe55Ni45,
foil, 25mm disks, 0.1mm thickness
Expand
View Pricing
GF63765827
Iron-Nickel alloy, Fe55Ni45,
foil, 5m coil, 0.1mm thickness
Expand
View Pricing
GF41310122
Iron-Nickel alloy, Fe55Ni45,
foil, 2m coil, 0.1mm thickness
Expand
View Pricing