![Iohexol Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/426/099/aedfa498-3080-4dd7-99e1-30b9ff7bcd7c/640/aedfa498-3080-4dd7-99e1-30b9ff7bcd7c.jpg)
Iohexol Related Compound B
![Iohexol Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/426/099/aedfa498-3080-4dd7-99e1-30b9ff7bcd7c/640/aedfa498-3080-4dd7-99e1-30b9ff7bcd7c.jpg)
Synonym(s):
5-Amino-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide, 5-Amino-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
Empirical Formula (Hill Notation):
C14H18I3N3O6
CAS Number:
Molecular Weight:
705.02
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2