Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iodoacetamide
Linear Formula:
ICH
2
CONH
2
CAS Number:
144-48-9
Molecular Weight:
184.96
EC Number:
205-630-1
Products
Product Category
bioactive small molecules (1)
Brand
SAFC (1)
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
A3221
Iodoacetamide,
Single use vial of 56 mg
Expand
View Pricing
I1149
Iodoacetamide,
BioUltra
Expand
View Pricing
I6125
Iodoacetamide,
≥99% (NMR), crystalline
Expand
View Pricing
I5161
Iodoacetamide,
Expand
View Pricing