
Iodine

Synonym(s):
Iodine
Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-11 of 11
Compare
Product Number
Description
Pricing