Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Invar
®
- controlled expansion alloy
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 43
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF22785475
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.02 mm, size 200 × 200 mm, temper as rolled
Expand
View Pricing
GF86911072
Invar
®
- controlled expansion alloy,
sheet, Fe64%/Ni36%, thickness 25 mm, size 52 × 52 mm, temper annealed
Expand
View Pricing
GF89829242
Invar
®
- controlled expansion alloy,
sheet, Fe64%/Ni36%, thickness 2.5 mm, size 100 × 100 mm, temper annealed
Expand
View Pricing
GF94449213
Invar
®
- controlled expansion alloy,
sheet, thickness 25 mm, Fe64%/Ni36%, size 102 × 102 mm, temper annealed
Expand
View Pricing
GF03944392
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.25 mm, length 1 m, coil width 50 mm
Expand
View Pricing
GF74553161
Invar
®
- controlled expansion alloy,
rod, 6.0 mm diameter, Fe64%/Ni36%, length 200 mm
Expand
View Pricing
GF55968216
Invar
®
- controlled expansion alloy,
rod, Fe 64%/Ni 36%, 6.0 mm diameter, length 100 mm
Expand
View Pricing
GF05606030
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.1 mm, size 25 × 25 mm, temper annealed
Expand
View Pricing
GF07362996
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.5 mm, size 300 × 450 mm, temper annealed
Expand
View Pricing
GF09527766
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.25 mm, size 200 × 200 mm, temper annealed
Expand
View Pricing
GF16098936
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.25 mm, length 5 m, coil width 50 mm
Expand
View Pricing
GF66490702
Invar
®
- controlled expansion alloy,
foil, thickness 0.5 mm, Fe64%/Ni36%, temper annealed, diameter 6 mm
Expand
View Pricing
GF46800615
Invar
®
- controlled expansion alloy,
rod, Fe 64%/Ni 36%, 6.0 mm diameter, length 500 mm
Expand
View Pricing
GF04614296
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.18 mm, length 0.5 m, temper hard
Expand
View Pricing
GF10205873
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.25 mm, length 2 m, coil width 50 mm
Expand
View Pricing
GF12814488
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.18 mm, 10 mm diameter, temper hard
Expand
View Pricing
GF24735539
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.5 mm, size 600 × 900 mm, temper annealed
Expand
View Pricing
GF38657571
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.18 mm, size 50 × 50 mm, temper hard
Expand
View Pricing
GF55236045
Invar
®
- controlled expansion alloy,
rod, Fe 64%/Ni 36%, 6.35 mm diameter, length 400 mm
Expand
View Pricing
GF55528207
Invar
®
- controlled expansion alloy,
foil, Fe 64%/Ni 36%, thickness 0.18 mm, size 200 × 200 mm, temper hard
Expand
View Pricing
Page 1 of 3
Page 1 of 3