Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inserts for 2 mL large opening vials, 6.0 mm I.D.
Products
Product Category
inserts (6)
Brand
Supelco (6)
ID (mm)
OD (mm)
Volume (ml)
Color
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
24715
Inserts for 2 mL large opening vials, 6.0 mm I.D.,
volume 0.35 mL, clear glass (shell style), O.D. × H × I.D. 6 mm × 31 mm × 4.6 mm, pkg of 100 ea
Expand
View Pricing
24721
Inserts for 2 mL large opening vials, 6.0 mm I.D.,
volume 0.2 mL, conical clear glass (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
Expand
View Pricing
24722
Inserts for 2 mL large opening vials, 6.0 mm I.D.,
volume 0.2 mL, conical polypropylene (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
Expand
View Pricing
24717
Inserts for 2 mL large opening vials, 6.0 mm I.D.,
volume 0.25 mL, conical clear glass, O.D. × H × I.D. 6 mm × 31 mm × 4.6 mm, pkg of 100 ea
Expand
View Pricing
24716
Inserts for 2 mL large opening vials, 6.0 mm I.D.,
volume 0.35 mL, clear glass (shell style), O.D. × H × I.D. 6 mm × 31 mm × 4.6 mm, pkg of 1000 ea
Expand
View Pricing
27407
Inserts for 2 mL large opening vials, 6.0 mm I.D.,
volume 0.25 mL, glass (with glass flange), for use with Step Vials, pkg of 100 ea
Expand
View Pricing