
Indomethacin Related Compound B

Synonym(s):
4-Chlorobenzoic acid
Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing