Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indium tin oxide
Synonym(s):
ITO
CAS Number:
50926-11-9
Products
Brand
Sigma-Aldrich (3)
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
544876
Indium tin oxide,
nanopowder, <50 nm particle size
Expand
View Pricing
747939
Indium tin oxide,
≥99.5% trace metals basis, 18 nm particle size (SEM), 20 wt. % in H
2
O
Expand
View Pricing
494682
Indium tin oxide,
−325 mesh, ≥99.99% trace metals basis
Expand
View Pricing