Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 61
Page 1 of 4
Page 1 of 4
Compare
Product Number
Description
Pricing
GF11368278
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
5m wire, 1.2mm diameter, annealed
Expand
View Pricing
GF08954792
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 200mm x 20mm diameter, annealed
Expand
View Pricing
GF48988782
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
5m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF58302277
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
foil, 0.5m coil, 0.25mm thickness, annealed
Expand
View Pricing
GF72084236
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
foil, 150 x 150mm, 0.1mm thickness, annealed
Expand
View Pricing
GF19399090
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 500mm x 20mm diameter, annealed
Expand
View Pricing
GF46916039
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
foil, 0.2m coil, 0.1mm thickness, annealed
Expand
View Pricing
GF10365779
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 100mm x 12.7mm diameter, annealed
Expand
View Pricing
GF88320203
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 500mm x 25.4mm diameter, annealed
Expand
View Pricing
GF98864364
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 100mm x 16mm diameter, annealed
Expand
View Pricing
GF88771012
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
50m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF41972531
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
20m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF34752719
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
foil, 300 x 300mm, 0.1mm thickness, annealed
Expand
View Pricing
GF33089418
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
foil, 100 x 100mm, 2.0mm thickness, annealed
Expand
View Pricing
GF59974778
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 200mm x 16mm diameter, annealed
Expand
View Pricing
GF43109917
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 200mm x 6.35mm diameter, annealed
Expand
View Pricing
GF33469533
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 100mm x 25.4mm diameter, annealed
Expand
View Pricing
GF82220330
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
rod, 100mm x 19mm diameter, annealed
Expand
View Pricing
GF44309909
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF24874803
Inconel
®
alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi,
foil, 50 x 50mm, 0.25mm thickness, annealed
Expand
View Pricing
Page 1 of 4
Page 1 of 4