Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
inconel
®
617
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF29909313
inconel
®
617,
rod, Ni57%/Cr22%/Co12%/Mo 9, 2.4 mm diameter, length 450 mm, temper annealed
Expand
View Pricing
GF82810223
inconel
®
617,
rod, Ni57%/Cr22%/Co12%/Mo 9%, 1.6 mm diameter, length 900 mm, temper annealed
Expand
View Pricing
GF45519004
inconel
®
617,
rod, Ni57%/Cr22%/Co12%/Mo 9, 2.4 mm diameter, length 900 mm, temper annealed
Expand
View Pricing
GF60386698
inconel
®
617,
rod, Ni57%/Cr22%/Co12%/Mo 9%, 19 mm diameter, length 50 mm, temper annealed
Expand
View Pricing
GF75177153
inconel
®
617,
rod, Ni57%/Cr22%/Co12%/Mo 9%, 1.6 mm diameter, length 450 mm, temper annealed
Expand
View Pricing