
Ibuprofen Ethyl Ester

Synonym(s):
Ethyl 2-(4-isobutylphenyl)propionate
Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing