Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hyoscine butylbromide for system suitability
Synonym(s):
(−)-Scopolamine N-butyl bromide, (−)-N-Butylscopolamine bromide, Hyoscine N-butyl bromide
Empirical Formula (Hill Notation):
C
21
H
30
BrNO
4
CAS Number:
149-64-4
Molecular Weight:
440.37
EC Number:
205-744-1
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
Y0001983
Hyoscine butylbromide for system suitability,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing