Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydroxyapatite
Synonym(s):
Calcium phosphate hydroxide, Durapatite, Hydroxylapatite
Linear Formula:
3Ca
3
(PO
4
)
2
· Ca(OH)
2
CAS Number:
1306-06-5
Molecular Weight:
502.31
EC Number:
215-145-7
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Special Grade
Puriss (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
21223
Hydroxyapatite,
purum p.a., ≥90% (as Ca
3
(PO
4
)
2
, KT)
Expand
View Pricing
04238
Hydroxyapatite,
puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Expand
View Pricing