
Human Insulin

Synonym(s):
Insulin human
Empirical Formula (Hill Notation):
C257H383N65O77S6
CAS Number:
Molecular Weight:
5807.57
Products
Brand
Formula Weight
Agency
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing