Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hirschmann
®
graduated pipette, Class AS
Products
Product Category
pipettes (7)
Color
Manufacturer
Capacity (mL)
Height (mm)
Glassware Class
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z610178
Hirschmann
®
graduated pipette, Class AS,
capacity 10 mL , 0.05 mL accuracy
Expand
View Pricing
Z610151
Hirschmann
®
graduated pipette, Class AS,
capacity 5 mL , 0.03 mL accuracy
Expand
View Pricing
Z610194
Hirschmann
®
graduated pipette, Class AS,
capacity 25 mL , 0.1 mL accuracy
Expand
View Pricing
Z610127
Hirschmann
®
graduated pipette, Class AS,
capacity 1 mL , 0.007 mL accuracy
Expand
View Pricing
Z610208
Hirschmann
®
graduated pipette, Class AS,
capacity 50 mL , 0.2 mL accuracy
Expand
View Pricing
Z610186
Hirschmann
®
graduated pipette, Class AS,
capacity 20 mL , 0.1 mL accuracy
Expand
View Pricing
Z610135
Hirschmann
®
graduated pipette, Class AS,
capacity 2 mL , 0.01 mL accuracy
Expand
View Pricing