Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified
Products
Manufacturer
Format
Application
Reference Material Type
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
S30000S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 4965 mPa.s (60 °C)
Expand
View Pricing
N600
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 149.2 mPa.s (60 °C)
Expand
View Pricing
S6S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 2.935 mPa.s (60 °C)
Expand
View Pricing
S60S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 20.05 mPa.s (60 °C)
Expand
View Pricing
S2000S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 431.7 mPa.s (60 °C)
Expand
View Pricing
S200S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 57.98 mPa.s (60 °C)
Expand
View Pricing
S3S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 1.605 mPa.s (60 °C)
Expand
View Pricing
S8000S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 1533 mPa.s (60 °C)
Expand
View Pricing
S20S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 7.806 mPa.s (60 °C)
Expand
View Pricing
N2000
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 431.7 mPa.s (60 °C)
Expand
View Pricing
N100S
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified,
viscosity 34.33 mPa.s (60 °C)
Expand
View Pricing